CHIS5-03371

5-03371 5-03372
76224-728EA 234.2 USD
76224-728 76224-726
CHIS5-03371
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR