CHIS5-03363

5-03363 5-03364
76224-710EA 65.51 USD
76224-710 76224-712
CHIS5-03363
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR