CHIS5-03357

5-03357 5-03358
76224-700EA 622.33 USD
76224-700 76224-698
CHIS5-03357
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR