CHIS5-03355

5-03356 5-03355
76224-696EA 646.89 USD
76224-696 76224-694
CHIS5-03355
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR