CHIS5-03347

5-03348 5-03347
76224-678EA 114.64 USD
76224-678 76224-680
CHIS5-03347
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR