CHIS5-03281

5-03282 5-03281
76224-578EA 81.89 USD
76224-578 76224-580
CHIS5-03281
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR