CHIS5-03265

5-03266 5-03265
76224-546EA 45.86 USD
76224-546 76224-548
CHIS5-03265
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR