CHIS5-03215

5-03215 5-03216
76224-478EA 65.51 USD
76224-478 76224-480
CHIS5-03215
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR