CHIS5-03201

5-03202 5-03201
76224-452EA 357.02 USD
76224-452 76224-450
CHIS5-03201
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR