CHIS5-03191

5-03192 5-03191
76224-432EA 319.36 USD
76224-432 76224-430
CHIS5-03191
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR