CHIS5-03187

5-03187 5-03188
76224-424EA 83.53 USD
76224-424 76224-422
CHIS5-03187
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR