CHIS5-03185

5-03185 5-03186
76224-420EA 327.54 USD
76224-420 76224-418
CHIS5-03185
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR