CHIS5-03179

5-03180 5-03179
76224-408EA 258.76 USD
76224-408 76224-406
CHIS5-03179
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR