CHIS5-03167

5-03167 5-03168
76225-860EA 235.83 USD
76225-860 76225-858
CHIS5-03167
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR