CHIS5-03163

5-03164 5-03163
76224-384EA 324.26 USD
76224-384 76224-382
CHIS5-03163
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR