CHIS5-03159

5-03160 5-03159
76224-376EA 221.09 USD
76224-376 76224-374
CHIS5-03159
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR