CHIS5-03129

5-03130 5-03129
76224-314EA 229.28 USD
76224-314 76224-316
CHIS5-03129
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR