CHIS5-02901

5-02901 5-02902
76223-914EA 311.16 USD
76223-914 76223-916
CHIS5-02901
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR