CHIS5-03061

5-03062 5-03061
76224-196EA 1072.7 USD
76224-196 76224-194
CHIS5-03061
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR