CHIS5-03029

5-03030 5-03029
76224-138EA 45.86 USD
76224-138 76224-140
CHIS5-03029
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR