CHIS5-03017

5-03017 5-03018
76224-116EA 265.31 USD
76224-116 76224-114
CHIS5-03017
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR