CHIS5-03011

5-03012 5-03011
76224-104EA 221.09 USD
76224-104 76224-102
CHIS5-03011
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR