CHIS5-03003

5-03004 5-03003
76224-088EA 900.74 USD
76224-088 76224-086
CHIS5-03003
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR