CHIS5-02997

5-02997 5-02998
76224-076EA 434 USD
76224-076 76224-074
CHIS5-02997
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR