CHIS5-02941

5-02941 5-02942
76223-978EA 131.02 USD
76223-978 76223-980
CHIS5-02941
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR