CHIS5-02907

5-02908 5-02907
76223-926EA 212.9 USD
76223-926 76223-928
CHIS5-02907
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR