CHIS5-02887

5-02888 5-02887
76223-886EA 94.99 USD
76223-886 76223-888
CHIS5-02887
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR