CHIS5-02799

5-02800 5-02799
76223-718EA 163.77 USD
76223-718 76223-720
CHIS5-02799
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR