CHIS5-02755

5-02755 5-02756
76223-648EA 491.31 USD
76223-648 76223-646
CHIS5-02755
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR