CHIS5-02751

5-02751 5-02752
76223-638EA 294.79 USD
76223-638 76223-640
CHIS5-02751
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR