CHIS5-02725

5-02725 5-02726
76223-586EA 162.14 USD
76223-586 76223-588
CHIS5-02725
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR