CHIS5-02711

5-02712 5-02711
76222-346EA 221.09 USD
76222-346 76222-348
CHIS5-02711
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR