CHIS5-02703

5-02703 5-02704
76222-330EA 209.63 USD
76222-330 76222-332
CHIS5-02703
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR