CHIS5-02691

5-02691 5-02692
76223-550EA 45.86 USD
76223-550 76223-552
CHIS5-02691
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR