CHIS5-02619

5-02619 5-02620
76223-430EA 81.89 USD
76223-430 76223-432
CHIS5-02619
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR