CHIS5-02617

5-02618 5-02617
76223-428EA 573.2 USD
76223-428 76223-426
CHIS5-02617
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR