CHIS5-02615

5-02616 5-02615
76223-422EA 163.77 USD
76223-422 76223-424
CHIS5-02615
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR