CHIS5-02607

5-02608 5-02607
76223-406EA 86.8 USD
76223-406 76223-408
CHIS5-02607
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR