CHIS5-02597

5-02598 5-02597
76223-386EA 98.26 USD
76223-386 76223-388
CHIS5-02597
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR