CHIS5-02589

5-02590 5-02589
76223-370EA 49.13 USD
76223-370 76223-372
CHIS5-02589
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR