CHIS5-02547

5-02548 5-02547
76223-302EA 262.03 USD
76223-302 76223-304
CHIS5-02547
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR