CHIS5-02519

5-02520 5-02519
76223-246EA 283.33 USD
76223-246 76223-248
CHIS5-02519
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR