CHIS5-02505

5-02505 5-02506
76223-234EA 360.29 USD
76223-234 76223-236
CHIS5-02505
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR