CHIS5-02493

5-02493 5-02494
76223-210EA 57.32 USD
76223-210 76223-212
CHIS5-02493
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR