CHIS5-02483

5-02484 5-02483
76223-192EA 157.22 USD
76223-192 76223-190
CHIS5-02483
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR