CHIS5-02439

5-02439 5-02440
76223-102EA 129.38 USD
76223-102 76223-104
CHIS5-02439
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR