CHIS5-02435

5-02435 5-02436
76223-094EA 114.64 USD
76223-094 76223-096
CHIS5-02435
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR