CHIS5-02431

5-02431 5-02432
76223-086EA 257.11 USD
76223-086 76223-088
CHIS5-02431
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR