CHIS5-02429

5-02429 5-02430
76223-082EA 90.08 USD
76223-082 76223-084
CHIS5-02429
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR