CHIS5-02423

5-02424 5-02423
76225-786EA 196.52 USD
76225-786 76225-788
CHIS5-02423
Reconstitution instructions: As indicated on manual
Storage conditions: Store at -20° C prior to opening
Shipping temperature: Room Temp
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR